
Benzyl alcohol 98.0-100.5% NF, Multi-Compendial low in peroxide
Supplier: Avantor
Certificates
About this item
CAS Number:100-51-6
:
Formula:C₆H₅CH₂OH
MDL Number:MFCD00004599
:108.14 g/mol
Density:1.05 g/cm³
:Management of Change (MOC) category = R
Flash Pt:100.40 °C
:GHS07
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- NF - Assay:98.0 - 100.5 %
- NF - Clarity of Solution:Passes Test
- NF - Color of Solution:Passes Test
- NF - Identification:Passes Test
- NF - Peroxide Value:≤5
- NF - Related Substances - Benzaldehyde:≤0.05 %
- NF - Related Substances - Cyclohexylmethanol:≤0.10 %
- NF - Related Substances - Total Other Peaks≤0.02 %
- NF - Related Substances - Total other peaks >RT of benzyl alcohol:≤0.2 %
- NF - Refractive Index, n²ºD:1.538 - 1.541
- NF - Acidity:Passes Test
- NF - Residue on Evaporation:≤0.05 %
- Solubility:Passes Test
- EP/BP - Assay:98.0 - 100.5 %
- EP/BP - Identification:Passes Test
- EP/BP - Acidity:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Refractive Index, n²ºD:1.538 - 1.541
- EP/BP - Peroxide Value:≤5
- EP/BP - Residue on Evaporation:≤0.05 %
- EP/BP - Related Substances - Benzaldehyde:≤0.05 %
- EP/BP - Related Substances - Cyclohexylmethanol:≤0.10 %
- EP/BP - Related Substances - Total Other Peaks≤0.02 %
- EP/BP - Related Substances - Total other peaks >RT of benzyl alcohol:≤0.2 %
- JP - Assay:98.0 - 100.5 %
- JP - Identification:Passes Test
- JP - Refractive Index, n²ºD:1.538 - 1.541
- JP - Specific Gravity at 20°/20°C:1.043 - 1.049
- JP - Clarity and Color of Solution:Passes Test
- JP - Acidity:Passes Test
- JP - Related Substances - Benzaldehyde:≤0.05 %
- JP - Related Substances - Cyclohexylmethanol:≤0.10 %
- JP - Related Substances - Total Other Peaks≤0.02 %
- JP - Related Substances - Total other peaks >RT of benzyl alcohol:≤0.2 %
- JP - Peroxide Value:≤5
- JP - Residue on Evaporation:≤0.05 %
- Total Aerobic Microbial Count:≤100 cfu/g
- Total Yeast and Mold Count:≤10 cfu/g
- Endotoxin Concentration, EU/mL:≤2.5