Order Entry
China
ContactUsLinkComponent
Kimtech™ G3 White Nitrile Gloves
Kimtech™ G3 White Nitrile Gloves
Supplier:  KIMBERLY CLARK
Certificates
undefined
Kimtech™ G3 White Nitrile Gloves
Supplier:  KIMBERLY CLARK
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Catalog No:
  • KIMB56882
  • KIMB56881
  • KIMB56882_U
  • KIMB56886
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Cleanroom class:
  • 1/ISO 3
  • 1/ISO 3
  • 1/ISO 3
  • 1/ISO 3
  • Finish:
  • Tacky finish, textured fingertips
  • Tacky finish, textured fingertips
  • Tacky finish, textured fingertips
  • Tacky finish, textured fingertips
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Cuff thickness:
  • 0,10 mil
  • 0,10 mil
  • 0,10 mil
  • 0,10 mil
  • Finger thickness:
  • 0,16 mil (middle finger)
  • 0,16 mil (middle finger)
  • 0,16 mil (middle finger)
  • 0,16 mil (middle finger)
  • Palm thickness:
  • 0,13 mil
  • 0,13 mil
  • 0,13 mil
  • 0,13 mil
  • Tensile strength:
  • 24 MPa, nominal (before aging)
  • 24 MPa, nominal (before aging)
  • 24 MPa, nominal (before aging)
  • 24 MPa, nominal (before aging)
  • EN norm:
  • EN ISO 374-1:2016 Type B (KPT) Chemical Splash protection; EN 374-4:2003 Resistance to degradation by chemicals; EN ISO 374-5:2016 Micro Organism and VIRUS Protection
  • EN ISO 374-1:2016 Type B (KPT) Chemical Splash protection; EN 374-4:2003 Resistance to degradation by chemicals; EN ISO 374-5:2016 Micro Organism and VIRUS Protection
  • EN ISO 374-1:2016 Type B (KPT) Chemical Splash protection; EN 374-4:2003 Resistance to degradation by chemicals; EN ISO 374-5:2016 Micro Organism and VIRUS Protection
  • EN ISO 374-1:2016 Type B (KPT) Chemical Splash protection; EN 374-4:2003 Resistance to degradation by chemicals; EN ISO 374-5:2016 Micro Organism and VIRUS Protection
  • Sterility:
  • Non sterile
  • Non sterile
  • Non sterile
  • Non sterile