Disposable tubing for use with bottles, Corning®
Chemically resistant, heat sealable flexible tubing. Accommodates both 2 L and 4 L bottles. Male MPC connector with cap.
- PureFit® tube clamp system
- Autoclavable
Chemically resistant, heat sealable flexible tubing. Accommodates both 2 L and 4 L bottles. Male MPC connector with cap.