Order Entry
China
ContactUsLinkComponent
Modified Impeller Assembly for Magnetically-Driven 3L Corning® Proculture® Spinner Flask
Modified Impeller Assembly for Magnetically-Driven 3L Corning® Proculture® Spinner Flask
Catalog # CORN402648
undefined
Modified Impeller Assembly for Magnetically-Driven 3L Corning® Proculture® Spinner Flask
Catalog # CORN402648

Specifications

  • For:
    3L angled sidearm spinner flasks (734-1912, 734-1918)
  • Description:
    Impeller assembly, stainless steel, dual bearing, for magnetically driven flask 3 L (to use with probes)
  • Seq:
    15
  • Pk:
    1

Specifications

About this item

Stainless steel impeller shaft with dual bearing, modified impeller assembly for use in magnetically driven, 3L Proculture® Spinner Flasks

  • Narrower impeller paddle design allows use with probes to create a small bioreactor
  • Non sterile
  • Reusable