Order Entry
China
Orders LinkContactUsLinkComponent
Kappaselect

Kappaselect

Affinity resin, KappaSelect, 25 mL
Catalog # GEHE17-5458-01
Supplier:   GE HEALTHCARE
Kappaselect
Catalog # GEHE17-5458-01
Supplier:  GE HEALTHCARE
CAS Number:  
Kappaselect

Affinity resin, KappaSelect, 25 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 25 毫升
Product Details & Documents

Antibody fragments, especially Fabs, are getting increased attention as potential biopharmaceuticals because they have some advantages over monoclonal antibodies (MAbs), KappaSelect is an affinity resin designed for the purification of human Fab (kappa) fragments.

Specifications

Specifications

  • Pk:
    25 mL
  • Description:
    Affinity resin, KappaSelect
  • Cat. No.:
    GEHE17-5458-01
  • Matrix:
    Highly cross-linked agarose
  • Ligand:
    Recombinant protein (Mr 13 000), produced in S. cerevisiae, with affinity for the constant domain of the immunoglobulin kappa light chain
  • pH range:
    3 – 10
  • Ligand density:
    ~5 mg/ml of medium
  • Storage temperature:
    2...8 °C

Specifications