


Acetonitrile ≥99.9%, B&J Brand™ LC-MS for LC-MS, for HPLC, Burdick & Jackson™
HONCLC015-4
:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
- GC Analysis:99.9 % min.
- Water by Karl Fischer Titration:0.01 % max.
- HPLC Gradient Elution:Passes test
- LC-MS Negative Mode (as 4-nitrophenol):50 ppb max.
- LC-MS Positive Mode (as caffeine):100 ppb max.
- LC-MS Positive Mode (as reserpine):50 ppb max.
- Refractive Index (20ºC):1.3434 - 1.3446
- Residue:1 mg/L max.
- Trace Metals:Passes test
- UV Cutoff:190 nm max.
:
:41.05 g/mol
:82 °C (1.013 hPa)
:–46 °C
:0.7822 g/cm³ (20 °C)
:6 °C
:Ambient
:200-835-2
:1648
:3,II
:GHS07
:GHS08
:GHS02
:75-05-8
:H₃CCN
:MFCD00001878