

Dichloromethane ≥99.9%, B&J Brand™ for HPLC, GC, pesticide residue analysis and spectrophotometry, Burdick & Jackson™
Catalog # HONC299-4
Supplier: HONEYWELL
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Pk:4 L
- Seq:5162
Specifications
Specification Test Results
- Acidity (as HCl):1 mg/L max.
- Chloride:10 mg/L max.
- Electron Capture GC:10 ng/L max.
- Flame Ionization GC (as nonane):5 ppb max.
- GC Analysis:99.9 % min.
- Refractive Index (20ºC):1.4236 - 1.4246
- Residue:1 mg/L max.
- UV Cutoff:233 nm max.
- Water by Karl Fischer Titration:0.01 % max.
Specification Test Results
About this item
:
:84.93 g/mol
Boiling Pt:40 °C
Melting Pt:–95 °C
Density:1.33 g/cm³
Flash Pt: Does not flash
Storage Temperature:Ambient
EINECS:200-838-9
UN:1593
ADR:6.1,III
:GHS07
:GHS08
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881