Order Entry
China
Orders LinkContactUsLinkComponent
Deferasirox, Reference Standards

Deferasirox, Reference Standards

Deferasirox, USP reference standard, Vial, 200 mg
Catalog # USPH1165656
Supplier:   U.S. PHARMACOPEIA
Deferasirox, Reference Standards
Catalog # USPH1165656
Supplier:  U.S. PHARMACOPEIA
CAS Number:  
Deferasirox, Reference Standards

Deferasirox, USP reference standard, Vial, 200 mg

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 200 毫克
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Deferasirox System Suitability Mixture: A mixture containing Deferasirox, Benzoxazinone analog, Deferasirox ethyl ester, Deferasirox 1,2-isomer, Deferasirox 1,3-isomer and Salicylimide

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    200 mg
  • Standards:
    USP reference standard
  • Description:
    Deferasirox
  • UN No.:
    3077
  • Cat. No.:
    USPH1165656
  • Description:
    Deferasirox
  • CAS No.:
    201530-41-8

Specifications