Fittings, Compression Nut Assembly with Ferrule
Supplier: MASTERFLEX PREMIUM
Certificates
About this item
Can handle pressures up to 225 psi.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rated to 220 psi maximum at 77 °F ( 25 °C). Usable from 32 to 212 °F (0 to 100 °C).
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLB31212-68
- MFLB31212-67
- MFLB31212-70
- MFLB31212-69
- MFLB31212-71
- MFLB31212-63
- MFLB31212-64
- MFLB31212-66
- MFLB31212-65
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year