Masterflex® Reducer Fittings, Hose Barb, Straight (HDPE), Avantor®
Best for low-pressure applications.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Temperture range −65 to +150 °F (−53 to +65 °C)
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
- Environmentally Preferable:
- Catalog No:
- MFLA30626-05
- MFLA30622-35
- MFLA30621-98
- MFLA30621-96
- MFLA41518-31
- MFLA30622-33
- MFLA30622-29
- MFLA30616-33
- MFLA30625-96
- MFLA30622-27
- MFLA30622-17
- MFLA30622-03
- MFLA30622-36
- MFLA30622-00
- MFLA30622-38
- MFLA30626-07
- MFLA30626-38
- MFLA30626-40
- MFLA31518-33
- MFLA30626-42
- MFLA30622-15
- MFLA30626-35
- MFLA30622-11
- MFLA30626-33
- MFLA30626-36
- MFLA30622-13
- MFLA30622-09
- MFLA30626-29
- MFLA30626-24
- MFLA30622-05
- MFLA30626-27
- MFLA30622-40
- MFLA30626-17
- MFLA30622-42
- MFLA30622-07
- MFLA30626-13
- MFLA41518-29
- MFLA41518-22
- MFLA30622-24
- MFLA30616-29
- Model:
- 30626-05
- 30622-35
- 30621-98
- 30621-96
- 41518-31
- 30622-33
- 30622-29
- 30616-33
- 30625-96
- 30622-27
- 30622-17
- 30622-03
- 30622-36
- 30622-00
- 30622-38
- 30626-07
- 30626-38
- 30626-40
- 31518-33
- 30626-42
- 30622-15
- 30626-35
- 30622-11
- 30626-33
- 30626-36
- 30622-13
- 30622-09
- 30626-29
- 30626-24
- 30622-05
- 30626-27
- 30622-40
- 30626-17
- 30622-42
- 30622-07
- 30626-13
- 41518-29
- 41518-22
- 30622-24
- 30616-29
- Sterilisation:
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene Oxide (ETO)
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene Oxide (ETO)
- Ethylene Oxide and autoclave
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only