Order Entry
China
ContactUsLinkComponent
Masterflex® Versatile Carboy Cap Adapter Inserts, Avantor®
Masterflex® Versatile Cap Adapter Insert, 83B, 3 x 1/4" ID Quick Connect Ports
Catalog # MFLZ06058-54
Supplier:  MASTERFLEX
Masterflex® Versatile Cap Adapter Insert, 83B, 3 x 1/4" ID Quick Connect Ports
Catalog # MFLZ06058-54
Supplier:  MASTERFLEX
Specifications
  • Cap material:
    PP
  • Cap Size:
    83B
  • Description:
    Versatile Cap Adapter Insert, 83B, Autoclavable, 3 x 1/4" ID Quick Connect Ports
  • Cap colour:
    White
  • Pk:
    1
Specifications
About this item
Interchangeable ports for flexibility.
  • Choose from a variety of sizes and configurations
  • Change the top connections of each carboy to match your application needs
  • Autoclavable
Versatile carboy cap adapter inserts are made of autoclavable polypropylene or clear polycarbonate. Quick-connect hosebarb fittings are also available. Vent ports are 1/4" in diameter.
  • Cap material:
    PP
  • Cap Size:
    83B
  • Description:
    Versatile Cap Adapter Insert, 83B, Autoclavable, 3 x 1/4" ID Quick Connect Ports
  • Environmentally Preferable:
  • Cap colour:
    White
  • Pk:
    1