Order Entry
China
ContactUsLinkComponent
Masterflex® Correlated Variable-Area Flowmeter with High and Low Optical Alarms, Avantor®
Masterflex® Correlated Variable-Area Flowmeter with High and Low Optical Alarms, Avantor®
Supplier:  MASTERFLEX PREMIUM
Certificates
undefined
Masterflex® Correlated Variable-Area Flowmeter with High and Low Optical Alarms, Avantor®
Supplier:  MASTERFLEX PREMIUM
Specifications
  • Model:
  • 33402-04
  • 33402-48
  • 33402-40
  • 33402-44
  • 33402-34
  • 33402-38
  • 33402-12
  • 33402-14
  • 33402-10
  • Float material:
  • Stainless Steel
  • Stainless Steel
  • Stainless Steel
  • Stainless Steel
  • Stainless Steel
  • Glass
  • Glass
  • Stainless Steel
  • Stainless Steel
  • Max. air flow rate:
  • 264 ml/min
  • 45227 ml/min
  • 4662 ml/min
  • 16737 ml/min
  • 814 ml/min
  • 2313 ml/min
  • 2313 ml/min
  • 4662 ml/min
  • 1682 ml/min
  • Repeatability:
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Specifications