Adapter Fittings, Compression to NPT(M), Brass
Create a secure connection with compression fittings.
- Constructed of durable brass
Good chemical resistance. Rated to 400 psi maximum at 77 °F ( 25 °C). Usable from -85 to 482 °F (-65 to 250 °C).
- Environmentally Preferable:
- Catalog No:
- MFLB31412-75
- MFLB31412-72
- MFLB31412-78
- MFLB31412-80
- MFLB31412-35
- MFLB31412-36
- MFLB31412-71
- MFLB31412-69
- Warranty:
- LTD life time
- LTD life time
- LTD life time
- LTD life time
- LTD life time
- LTD life time
- LTD life time
- LTD life time