Order Entry
China
ContactUsLinkComponent
Venture Tech Sanitary Clamp Ferrules, PFA
Venture Tech Sanitary Clamp Ferrules, PFA
供应商:  MASTERFLEX PREMIUM
证书
undefined
Venture Tech Sanitary Clamp Ferrules, PFA
供应商:  MASTERFLEX PREMIUM
规格
  • Max. operating pressure:
  • 100 psi in ambient conditions; 60 psi at 180 °F (82 °C); 60 psi at 35 °F (2 °C)
  • 100 psi in ambient conditions; 60 psi at 180 °F (82 °C); 60 psi at 35 °F (2 °C)
  • 100 psi in ambient conditions; 60 psi at 180 °F (82 °C); 60 psi at 35 °F (2 °C)
  • Max. temperature:
  • 82 °C at 1 bar (180 °F at 14,7 psig)
  • 82 °C at 1 bar (180 °F at 14,7 psig)
  • 82 °C at 1 bar (180 °F at 14,7 psig)
  • Max. vacuum:
  • 812 mm Hg (32" Hg)
  • 812 mm Hg (32" Hg)
  • 812 mm Hg (32" Hg)
  • Sterilisation:
  • Autoclave only
  • Autoclave only
  • Autoclave only
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
规格