Order Entry
China
Orders LinkContactUsLinkComponent
Masterflex® Adapter Fittings, Male Luer to Hose Barb, Straight, Polycarbonate, Avantor®
Masterflex® Adapter Fittings, Male Luer to Hose Barb, Straight, Polycarbonate, Avantor®
Masterflex® Adapter Fittings, Male Luer to Hose Barb, Straight, Polycarbonate, Avantor®
Masterflex® Adapter Fittings, Male Luer to Hose Barb, Straight, Polycarbonate, Avantor®
Ensure your process integrity with full material traceability.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Barbed adapters feature three styles of barbs: standard, wide-bore, and low-profile. Wide-bore barbs offer enhanced fluid flow through the fitting and are usable with any flexible tubing. Low-profile barbs are ideal for use with semi-rigid tubing, such as nylon, polyethylene, or polypropylene. Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity.
Hose barbs are available from ¹/₁₆" to ¹/₄" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.
  • Connection type (port 1):
  • Male luer lock
  • Male luer lock
  • Male luer lock
  • Male luer lock
  • Male luer lock
  • Male luer lock
  • Male luer slip
  • No. of ports:
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • Warranty:
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
Product Family Options
  • Description
    Barb size/tubing int.Ø (port 1)
    Connection Type
    Fitting material
    Flow pattern
  • Fitting, Polycarbonate, Straight, Male Luer Lock to Hosebarb Adapter, 1/16" ID; 25/PK
    1/16"
    Male Luer to Barb
    Polycarbonate
    Straight
    1 * 25 件
    1 * 25 件
    Avantor exclusive
    CE certificate
    Specifications
    Cat. No.
    MFLA45504-00
    Connection type (port 1)
    Male luer lock
    No. of ports
    2
    Warranty
    3 Months
    Avantor exclusive
    CE certificate
  • Fitting, Polycarbonate, Straight, Male Luer Lock to Hosebarb Adapter, 3/32" ID; 25/PK
    3/32"
    Male Luer to Barb
    Polycarbonate
    Straight
    1 * 25 件
    1 * 25 件
    Avantor exclusive
    CE certificate
    Specifications
    Cat. No.
    MFLA45504-02
    Connection type (port 1)
    Male luer lock
    No. of ports
    2
    Warranty
    3 Months
    Avantor exclusive
    CE certificate
  • Fitting, Polycarbonate, Straight, Male Luer Lock to Hosebarb Adapter, 1/8" ID; 25/PK
    1/8"
    Male Luer to Barb
    Polycarbonate
    Straight
    1 * 25 件
    1 * 25 件
    Avantor exclusive
    CE certificate
    Specifications
    Cat. No.
    MFLA45504-04
    Connection type (port 1)
    Male luer lock
    No. of ports
    2
    Warranty
    3 Months
    Avantor exclusive
    CE certificate
  • Fitting, Polycarbonate, Straight, Male Luer Lock to Hosebarb Adapter, 5/32" ID; 25/PK
    5/32"
    Male Luer to Barb
    Polycarbonate
    Straight
    1 * 25 件
    1 * 25 件
    Avantor exclusive
    CE certificate
    Specifications
    Cat. No.
    MFLA45504-06
    Connection type (port 1)
    Male luer lock
    No. of ports
    2
    Warranty
    3 Months
    Avantor exclusive
    CE certificate
  • Fitting, Polycarbonate, Straight, Male Luer Lock to Hosebarb Adapter, 3/16" ID; 25/PK
    3/16"
    Male Luer to Barb
    Polycarbonate
    Straight
    1 * 25 件
    1 * 25 件
    Avantor exclusive
    CE certificate
    Specifications
    Cat. No.
    MFLA45504-08
    Connection type (port 1)
    Male luer lock
    No. of ports
    2
    Warranty
    3 Months
    Avantor exclusive
    CE certificate
  • Fitting, Polycarbonate, Straight, Male Luer Lock to Hosebarb Adapter, 1/4" ID; 25/PK
    1/4"
    Male Luer to Barb
    Polycarbonate
    Straight
    1 * 25 件
    1 * 25 件
    Avantor exclusive
    CE certificate
    Specifications
    Cat. No.
    MFLA45504-19
    Connection type (port 1)
    Male luer lock
    No. of ports
    2
    Warranty
    3 Months
    Avantor exclusive
    CE certificate
  • Fitting, Polycarbonate, Straight, Male Slip Luer with Separate Lock Ring to Hose Barb Adapter, 1/4" ID; 25/PK
    1/4"
    Male Luer to Barb
    Polycarbonate
    Straight
    1 * 25 件
    1 * 25 件
    Avantor exclusive
    CE certificate
    Specifications
    Cat. No.
    MFLA45504-50
    Connection type (port 1)
    Male luer slip
    No. of ports
    2
    Warranty
    3 Months
    Avantor exclusive
    CE certificate