Order Entry
China
ContactUsLinkComponent
Masterflex® Sanitary Fitting Clamps, PVDF, Avantor®
Masterflex® Sanitary Fitting Clamps, PVDF, Avantor®
Certificates
undefined
Masterflex® Sanitary Fitting Clamps, PVDF, Avantor®
Specifications
  • Model:
  • 31811-01
  • 31811-02
  • 31811-00
  • Sanitary clamp size (port 1):
  • 1-1/2"
  • 2"
  • 3/4"
  • Connection type:
  • Sanitary
  • Sanitary
  • Sanitary
  • Chemical resistance:
  • Very good
  • Very good
  • Very good
  • Min. temperature:
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • Max. temperature:
  • 138 °C (280 °F)
  • 138 °C (280 °F)
  • 138 °C (280 °F)
  • Max. operating pressure:
  • 150 psi
  • 150 psi
  • 150 psi
  • Sterilisation:
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
Specifications