24-well deep well plates, rectangular wells, Axygen®
Catalog # AXYGP-DW-10ML-24-C
Supplier: Corning
Specifications
- Description:Deep well plates
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Pk:25
- Well volume:10 ml
Specifications
About this item
Axygen® deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage.
Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to −80°C. Ultra-flat surface ensures proper sealing with heat sealing film. Clear, V shaped well bottom.