PCR microplates, 384-well Rigi-Plate, Axygen®
Catalog # AXYGPCR-384-RGD-BK
Supplier: Corning
Specifications
- No. of wells:384
- Skirt:Fully skirted
- Well colour:Black
- Pk:20
- Well volume:0,04 ml
Specifications
About this item
Flat bottom, deep well microplates are designed to provide extra rigidity in thermal cycling applications.
Constructed of sturdy and durable polypropylene, plates are available in three color styles for application specific procedures. Choice of clear, black, or white plates allow for colorimetric, fluorescent, or luminescent applications. Certified RNase-, DNase-, and pyrogen-free. Well volume: 40 µl.
Ordering information: Clear plates are packaged 20 plates per pack, 5 packs per case. Black and white plates are packaged in sleeves of 20, with 5 sleeves per case.