Order Entry
China
ContactUsLinkComponent
FMI Valveless Piston Pump Heads, Series Q
FMI Valveless Piston Pump Heads, Series Q
Certificates
undefined
FMI Valveless Piston Pump Heads, Series Q
Specifications
  • Accuracy:
  • ±0,1%
  • ±0,1%
  • ±0,1%
  • ±0,1%
  • ±0,1%
  • ±0,1%
  • ±0,1%
  • ±0,1%
  • Duty cycle continuous:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Inlet connection:
  • 1/4" compression
  • 1/4" compression
  • 1/4" compression
  • 0,25 NPT(F)
  • 0,25 NPT(F)
  • 0,25 NPT(F)
  • Max. pressure:
  • 4,1 bar (60 psi)
  • 4,1 bar (60 psi)
  • 4,1 bar (60 psi)
  • 4,1 bar (60 psi)
  • 4,1 bar (60 psi)
  • 6,9 bar (100 psi)
  • 6,9 bar (100 psi)
  • 4,1 bar (100 psi)
  • Max. temperature:
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 177 °C (350 °F)
  • 177 °C (350 °F)
  • 177 °C (350 °F)
  • Max. viscosity:
  • 500 cP
  • 500 cP
  • 500 cP
  • 500 cP
  • 500 cP
  • 500 cP
  • 500 cP
  • 500 cP
  • Outlet connection:
  • 1/4" compression
  • 1/4" compression
  • 1/4" compression
  • 0,25 NPT(F)
  • 0,25 NPT(F)
  • 0,25 NPT(F)
  • Suction lift, wet:
  • 4572 mm of H₂O (180" H₂O)
  • 4572 mm of H₂O (180" H₂O)
  • 4572 mm of H₂O (180" H₂O)
  • 4572 mm of H₂O (180" H₂O)
  • 4572 mm of H₂O (180" H₂O)
  • 4572 mm of H₂O (180" H₂O)
  • 4572 mm of H₂O (180" H₂O)
  • 4572 mm of H₂O (180" H₂O)
  • Wetted materials:
  • Ceramic Piston, PVDF Cylinder Body, Carbon Cylinder Liner
  • Ceramic Piston and Cylinder Liner, PVDF Cylinder Body
  • Ceramic Piston, Stainless Steel Cylinder Body, Ceramic Cylinder Liner
  • Ceramic Piston and Cylinder Liner, PVDF Cylinder Body
  • Ceramic Piston, PVDF Cylinder Body, Ceramic Cylinder Liner
  • Ceramic Piston and Cylinder Liner, Stainless Steel Cylinder Body
  • Ceramic Piston, Stainless Steel Cylinder Body, Carbon Cylinder Liner
  • Ceramic Piston and Cylinder Liner, Stainless Steel Cylinder Body
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Specifications