Order Entry
China
Orders LinkContactUsLinkComponent
J.T.Baker®, Syringe Filters
 :   Avantor
J.T.Baker®, Syringe Filters
 :  Avantor
J.T.Baker®, Syringe Filters
J.T.Baker®, Syringe Filters
These syringe filters are specifically designed for chromatography sample preparation applications and optimised to provide the most consistent results with less extractables.
  • HPLC certified
  • Biosafe according to Class VI plastics tests
  • Polypropylene housing material
  • Non sterile
  • Each filter has a female Luer-lock inlet and a male slip Luer outlet
  • Large pore size 1,0 µm for high throughput and fast flow rates
Available with PTFE (hydrophilic or hydrophobic), nylon, PES, RC membrane and GF (glass fibre):
- Water wettable hydrophilic PTFE offers excellent chemical resistance to both common aqueous and aggressive organic solvents, most of the acids and alkalis
- Hydrophobic PTFE resistant to the majority of solvents as well as strong acids and bases, also suitable for gas filtration
- Nylon provides a wide range of chemical compatibility, suitable for filtration of either aqueous or organic solvents. It is a natural hydrophilic membrane that can be used in a broad pH range
- PES offers lower protein binding as well as higher flow rates
- Regenerated cellulose exhibits low protein binding and wide chemical resistance
- Built in glass fibre (100% binder-free borosilicate glass fibre) pre-filter double-layer type is specially for high particle loading, ideal for difficult to filter liquids
 : Manufactured under ISO 9001:2015 quality management.
Product Family Options
Glass fibre (GF)
  • Ø
    Pore size
    Membrane
    Packed
  • 13 mm
    1,00 µm
    Glass fibre (GF)
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥448.00
    ¥448.00
    1 * 200 件
       ¥448.00
  • 13 mm
    1,00 µm
    Glass fibre (GF)
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥2,189.00
    ¥2,189.00
    1 * 1.000 件
       ¥2,189.00
  • 25 mm
    1,00 µm
    Glass fibre (GF)
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥637.00
    ¥637.00
    1 * 200 件
       ¥637.00
  • 25 mm
    1,00 µm
    Glass fibre (GF)
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥3,125.00
    ¥3,125.00
    1 * 1.000 件
       ¥3,125.00
  • 30 mm
    1,00 µm
    Glass fibre (GF)
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥708.00
    ¥708.00
    1 * 200 件
       ¥708.00
  • 30 mm
    1,00 µm
    Glass fibre (GF)
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥3,437.00
    ¥3,437.00
    1 * 1.000 件
       ¥3,437.00
Hydrophilic polytetrafluoroethylene (PTFE)
  • Ø
    Pore size
    Membrane
    Packed
  • 13 mm
    0,22 µm
    Hydrophilic PTFE
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥585.00
    ¥585.00
    1 * 200 件
       ¥585.00
  • 13 mm
    0,22 µm
    Hydrophilic PTFE
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥2,866.00
    ¥2,866.00
    1 * 1.000 件
       ¥2,866.00
  • 13 mm
    0,45 µm
    Hydrophilic PTFE
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥585.00
    ¥585.00
    1 * 200 件
       ¥585.00
  • 13 mm
    0,45 µm
    Hydrophilic PTFE
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥2,866.00
    ¥2,866.00
    1 * 1.000 件
       ¥2,866.00
  • 25 mm
    0,22 µm
    Hydrophilic PTFE
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥796.00
    ¥796.00
    1 * 200 件
       ¥796.00
  • 25 mm
    0,22 µm
    Hydrophilic PTFE
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥3,908.00
    ¥3,908.00
    1 * 1.000 件
       ¥3,908.00
  • 25 mm
    0,45 µm
    Hydrophilic PTFE
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥949.00
    ¥949.00
    1 * 200 件
       ¥949.00
  • 25 mm
    0,45 µm
    Hydrophilic PTFE
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥3,908.00
    ¥3,908.00
    1 * 1.000 件
       ¥3,908.00
  • 25 mm
    0,45 µm
    Hydrophilic PTFE, GF prefilter
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥952.00
    ¥952.00
    1 * 200 件
       ¥952.00
  • 25 mm
    0,45 µm
    Hydrophilic PTFE, GF prefilter
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥4,688.00
    ¥4,688.00
    1 * 1.000 件
       ¥4,688.00
  • 30 mm
    0,22 µm
    Hydrophilic PTFE
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥853.00
    ¥853.00
    1 * 200 件
       ¥853.00
  • 30 mm
    0,22 µm
    Hydrophilic PTFE
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥4,167.00
    ¥4,167.00
    1 * 1.000 件
       ¥4,167.00
  • 30 mm
    0,45 µm
    Hydrophilic PTFE
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥853.00
    ¥853.00
    1 * 200 件
       ¥853.00
  • 30 mm
    0,45 µm
    Hydrophilic PTFE
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥4,167.00
    ¥4,167.00
    1 * 1.000 件
       ¥4,167.00
Nylon
  • Ø
    Pore size
    Membrane
    Packed
  • 13 mm
    0,22 µm
    Nylon
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥418.00
    ¥418.00
    1 * 200 件
       ¥418.00
  • 13 mm
    0,22 µm
    Nylon
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥2,031.00
    ¥2,031.00
    1 * 1.000 件
       ¥2,031.00
  • 13 mm
    0,45 µm
    Nylon
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥418.00
    ¥418.00
    1 * 200 件
       ¥418.00
  • 13 mm
    0,45 µm
    Nylon
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥2,031.00
    ¥2,031.00
    1 * 1.000 件
       ¥2,031.00
  • 25 mm
    0,22 µm
    Nylon
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥475.00
    ¥475.00
    1 * 200 件
       ¥475.00
  • 25 mm
    0,20 µm
    Nylon
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥2,290.00
    ¥2,290.00
    1 * 1.000 件
       ¥2,290.00
  • 25 mm
    0,45 µm
    Nylon
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥473.00
    ¥473.00
    1 * 200 件
       ¥473.00
  • 25 mm
    0,45 µm
    Nylon
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥2,290.00
    ¥2,290.00
    1 * 1.000 件
       ¥2,290.00
  • 25 mm
    0,45 µm
    Nylon, GF prefilter
    2 jars of 100
    1 * 200 件
    1 * 200 件
  • 25 mm
    0,45 µm
    Nylon, GF prefilter
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥3,125.00
    ¥3,125.00
    1 * 1.000 件
       ¥3,125.00
  • 30 mm
    1,00 µm
    Nylon
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥532.00
    ¥532.00
    1 * 200 件
       ¥532.00
  • 30 mm
    1,00 µm
    Nylon
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥2,554.00
    ¥2,554.00
    1 * 1.000 件
       ¥2,554.00
  • 30 mm
    0,22 µm
    Nylon
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥532.00
    ¥532.00
    1 * 200 件
       ¥532.00
  • 30 mm
    0,22 µm
    Nylon
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥2,554.00
    ¥2,554.00
    1 * 1.000 件
       ¥2,554.00
  • 30 mm
    0,45 µm
    Nylon
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥532.00
    ¥532.00
    1 * 200 件
       ¥532.00
  • 30 mm
    0,45 µm
    Nylon
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥2,554.00
    ¥2,554.00
    1 * 1.000 件
       ¥2,554.00
Polyethersulfone (PES)
  • Ø
    Pore size
    Membrane
    Packed
  • 13 mm
    0,10 µm
    PES
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥479.00
    ¥479.00
    1 * 200 件
       ¥479.00
  • 13 mm
    0,10 µm
    PES
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥2,343.00
    ¥2,343.00
    1 * 1.000 件
       ¥2,343.00
  • 13 mm
    0,22 µm
    PES
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥479.00
    ¥479.00
    1 * 200 件
       ¥479.00
  • 13 mm
    0,22 µm
    PES
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥2,343.00
    ¥2,343.00
    1 * 1.000 件
       ¥2,343.00
  • 13 mm
    0,45 µm
    PES
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥481.00
    ¥481.00
    1 * 200 件
       ¥481.00
  • 13 mm
    0,45 µm
    PES
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥2,343.00
    ¥2,343.00
    1 * 1.000 件
       ¥2,343.00
  • 13 mm
    0,45 µm
    PES, GF prefilter
    2 jars of 100
    1 * 200 件
    1 * 200 件
  • 13 mm
    0,45 µm
    PES, GF prefilter
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥3,125.00
    ¥3,125.00
    1 * 1.000 件
       ¥3,125.00
  • 25 mm
    0,10 µm
    PES
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥637.00
    ¥637.00
    1 * 200 件
       ¥637.00
  • 25 mm
    0,10 µm
    PES
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥3,125.00
    ¥3,125.00
    1 * 1.000 件
       ¥3,125.00
  • 25 mm
    0,22 µm
    PES
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥637.00
    ¥637.00
    1 * 200 件
       ¥637.00
  • 25 mm
    0,22 µm
    PES
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥3,125.00
    ¥3,125.00
    1 * 1.000 件
       ¥3,125.00
  • 25 mm
    0,45 µm
    PES
    2 jars of 100
    1 * 200 件
    1 * 200 件
  • 25 mm
    0,45 µm
    PES
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥3,125.00
    ¥3,125.00
    1 * 1.000 件
       ¥3,125.00
  • 25 mm
    0,45 µm
    PES, GF prefilter
    2 jars of 100
    1 * 200 件
    1 * 200 件
  • 25 mm
    0,45 µm
    PES, GF prefilter
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥3,906.00
    ¥3,906.00
    1 * 1.000 件
       ¥3,906.00
  • 30 mm
    1,00 µm
    PES
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥699.00
    ¥699.00
    1 * 200 件
       ¥699.00
  • 30 mm
    1,00 µm
    PES
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥3,385.00
    ¥3,385.00
    1 * 1.000 件
       ¥3,385.00
  • 30 mm
    0,10 µm
    PES
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥699.00
    ¥699.00
    1 * 200 件
       ¥699.00
  • 30 mm
    0,10 µm
    PES
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥3,385.00
    ¥3,385.00
    1 * 1.000 件
       ¥3,385.00
  • 30 mm
    0,22 µm
    PES
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥699.00
    ¥699.00
    1 * 200 件
       ¥699.00
  • 30 mm
    0,22 µm
    PES
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥3,385.00
    ¥3,385.00
    1 * 1.000 件
       ¥3,385.00
  • 30 mm
    0,45 µm
    PES
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥698.00
    ¥698.00
    1 * 200 件
       ¥698.00
  • 30 mm
    0,45 µm
    PES
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥3,385.00
    ¥3,385.00
    1 * 1.000 件
       ¥3,385.00
  • 30 mm
    0,45 µm
    PES, GF prefilter
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥855.00
    ¥855.00
    1 * 200 件
       ¥855.00
  • 30 mm
    0,45 µm
    PES, GF prefilter
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥4,167.00
    ¥4,167.00
    1 * 1.000 件
       ¥4,167.00
Hydrophobic polytetrafluoroethylene (PTFE)
  • Ø
    Pore size
    Membrane
    Packed
  • 13 mm
    0,22 µm
    Hydrophobic PTFE
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥479.00
    ¥479.00
    1 * 200 件
       ¥479.00
  • 13 mm
    0,22 µm
    Hydrophobic PTFE
    Bulk
    1 * 1.000 件
    1 * 1.000 件
  • 13 mm
    0,45 µm
    Hydrophobic PTFE
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥479.00
    ¥479.00
    1 * 200 件
       ¥479.00
  • 13 mm
    0,45 µm
    Hydrophobic PTFE
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥2,343.00
    ¥2,343.00
    1 * 1.000 件
       ¥2,343.00
  • 25 mm
    0,22 µm
    Hydrophobic PTFE
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥637.00
    ¥637.00
    1 * 200 件
       ¥637.00
  • 25 mm
    0,22 µm
    Hydrophobic PTFE
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥3,125.00
    ¥3,125.00
    1 * 1.000 件
       ¥3,125.00
  • 25 mm
    0,45 µm
    Hydrophobic PTFE
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥639.00
    ¥639.00
    1 * 200 件
       ¥639.00
  • 25 mm
    0,45 µm
    Hydrophobic PTFE
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥3,125.00
    ¥3,125.00
    1 * 1.000 件
       ¥3,125.00
  • 25 mm
    0,45 µm
    Hydrophobic PTFE, GF prefilter
    2 jars of 100
    1 * 200 件
    1 * 200 件
  • 25 mm
    0,45 µm
    Hydrophobic PTFE, GF prefilter
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥3,906.00
    ¥3,906.00
    1 * 1.000 件
       ¥3,906.00
  • 30 mm
    1,00 µm
    Hydrophobic PTFE
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥699.00
    ¥699.00
    1 * 200 件
       ¥699.00
  • 30 mm
    1,00 µm
    Hydrophobic PTFE
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥3,385.00
    ¥3,385.00
    1 * 1.000 件
       ¥3,385.00
  • 30 mm
    0,22 µm
    Hydrophobic PTFE
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥699.00
    ¥699.00
    1 * 200 件
       ¥699.00
  • 30 mm
    0,22 µm
    Hydrophobic PTFE
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥3,385.00
    ¥3,385.00
    1 * 1.000 件
       ¥3,385.00
  • 30 mm
    0,45 µm
    Hydrophobic PTFE
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥699.00
    ¥699.00
    1 * 200 件
       ¥699.00
  • 30 mm
    0,45 µm
    Hydrophobic PTFE
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥3,385.00
    ¥3,385.00
    1 * 1.000 件
       ¥3,385.00
Regenerated cellulose (RC)
  • Ø
    Pore size
    Membrane
    Packed
  • 13 mm
    0,22 µm
    RC
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥1,209.00
    ¥1,209.00
    1 * 200 件
       ¥1,209.00
  • 13 mm
    0,22 µm
    RC
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥5,991.00
    ¥5,991.00
    1 * 1.000 件
       ¥5,991.00
  • 13 mm
    0,45 µm
    RC
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥1,209.00
    ¥1,209.00
    1 * 200 件
       ¥1,209.00
  • 13 mm
    0,45 µm
    RC
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥5,991.00
    ¥5,991.00
    1 * 1.000 件
       ¥5,991.00
  • 25 mm
    0,22 µm
    RC
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥1,367.00
    ¥1,367.00
    1 * 200 件
       ¥1,367.00
  • 25 mm
    0,22 µm
    RC
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥6,769.00
    ¥6,769.00
    1 * 1.000 件
       ¥6,769.00
  • 25 mm
    0,45 µm
    RC
    2 jars of 100
    1 * 200 件
    1 * 200 件
  • 25 mm
    0,45 µm
    RC
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥6,769.00
    ¥6,769.00
    1 * 1.000 件
       ¥6,769.00
  • 25 mm
    0,45 µm
    RC, GF prefilter
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥1,525.00
    ¥1,525.00
    1 * 200 件
       ¥1,525.00
  • 25 mm
    0,45 µm
    RC, GF prefilter
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥7,552.00
    ¥7,552.00
    1 * 1.000 件
       ¥7,552.00
  • 30 mm
    0,22 µm
    RC
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥1,442.00
    ¥1,442.00
    1 * 200 件
       ¥1,442.00
  • 30 mm
    0,22 µm
    RC
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥7,108.00
    ¥7,108.00
    1 * 1.000 件
       ¥7,108.00
  • 30 mm
    0,45 µm
    RC
    2 jars of 100
    1 * 200 件
    1 * 200 件 - ¥1,442.00
    ¥1,442.00
    1 * 200 件
       ¥1,442.00
  • 30 mm
    0,45 µm
    RC
    Bulk
    1 * 1.000 件
    1 * 1.000 件 - ¥7,108.00
    ¥7,108.00
    1 * 1.000 件
       ¥7,108.00