Syringe filters, prefilter, sterile, Whatman™ Anotop Plus
Supplier: CYTIVA WHATMAN
Certificates
About this item
Cytiva's Whatman Anotop Plus sterile syringe filter with glass microfiber prefilter is compatible with most organic and aqueous solutions for use in a wide variety of applications.
- Contains an inorganic Anopore membrane and is available in three pore sizes
- Polypropylene syringe filters comes in two diameters depending on sample volume
- Integrated glass microfiber prefilter aids in filtration of difficult samples
- Recyclable after use
Anotop Plus filters are manufactured with an Anopore membrane suitable for many applications, including capillary electrophoresis, UV/Vis sample preparation, protein separation, and HPLC sample preparation.
Specifications
- Catalog No:
- WHAT6809-3102
- WHAT6809-4102
- Type:
- Anotop™ 10 Plus
- Anotop™ 25 Plus
- Ø:
- 10 mm
- 25 mm
- Dimensions:
- 15,4×18,5 mm
- 36,8×26,3 mm
- Max. operating pressure:
- 6,9 bar
- 6,9 bar
- Pre-filter:
- Glass microfibre (binderless)
- Glass microfibre (binderless)