Anti-CD8a Mouse Monoclonal Antibody [clone: RPA-T8] (FITC (Fluorescein Isothiocyanate))
BLEG301050
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antigen symbol:CD8a
- Clonality:Monoclonal
- Clone:RPA-T8
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Baboon,Cynomolgus monkey,Sooty mangabey,Rhesus monkey,Pigtailed macaque,Chimpanzee,Human
- Antigen synonyms:T8|Leu2
- Storage buffer:µg size: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
test sizes: Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA). - Storage temperature:2…8 °C
- Tested applications:FC
- Purification:The antibody was purified by affinity chromatography, and conjugated with FITC under optimal conditions.
- Size:500 tests
- Pk:500 Tests