About this item
Phenyl Sepharose 6 Fast Flow is a well-established, standard aromatic HIC medium for capture and intermediate purification requiring low to medium hydrophobicity.
- Provide convenient and fast small-scale protein purifications.
- Phenyl Sepharose 6 Fast Flow is a well-established, standard aromatic HIC medium for capture and intermediate purification requiring low to medium hydrophobicity. Two levels of substitution on Phenyl Sepharose 6 Fast Flow help find optimal selectivity and binding capacity.
- Flexible design for use with syringes, single laboratory pump, or chromatography system such as ÄKTA design.
- High flow rates and high capacities.
- Excellent and easy scale-up achieved by connecting several HiTrap columns in series or using HiPrep columns.
Two levels of substitution on Phenyl Sepharose 6 Fast Flow help find optimal selectivity and binding capacity. HiTrap Phenyl FF (Low Sub) are prepacked Phenyl Sepharose 6 Fast Flow columns for hydrophobic interaction chromatography (HIC). The columns are used for small-scale protein purification as well as for screening of binding and elution conditions.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- GEHE17-5193-01
- GEHE17-5194-01
- Max. pressure:
- 5 bar
- 5 bar
- pH range:
- 3 - 13
- 3 - 13