HiTrap columns, Phenyl FF (High Sub)
About this item
Phenyl Sepharose 6 Fast Flow is a well-established, standard aromatic HIC medium for capture and intermediate purification requiring low to medium hydrophobicity.
- Provide convenient and fast small-scale protein purifications
- Two levels of substitution on Phenyl Sepahrose 6 Fast Flow help find optimal selectivity and binding capacity
- Flexible design for use with syringes, single laboratory pump, or chromatography system such as ÄKTA design
- High flow rates and high capacities
- Excellent and easy scale-up achieved by connecting several HiTrap columns in series or using HiPrep columns
HiTrap Phenyl FF (high sub) are prepacked Phenyl Sepharose 6 Fast Flow columns for hydrophobic interaction chromatography (HIC). The columns are used for small-scale protein purification as well as for screening of binding and elutions conditions.
2 Options Available Below
Product Details & Documents
Phenyl Sepharose 6 Fast Flow is a well-established, standard aromatic HIC medium for capture and intermediate purification requiring low to medium hydrophobicity.
- Provide convenient and fast small-scale protein purifications
- Two levels of substitution on Phenyl Sepahrose 6 Fast Flow help find optimal selectivity and binding capacity
- Flexible design for use with syringes, single laboratory pump, or chromatography system such as ÄKTA design
- High flow rates and high capacities
- Excellent and easy scale-up achieved by connecting several HiTrap columns in series or using HiPrep columns
HiTrap Phenyl FF (high sub) are prepacked Phenyl Sepharose 6 Fast Flow columns for hydrophobic interaction chromatography (HIC). The columns are used for small-scale protein purification as well as for screening of binding and elutions conditions.
Specifications for Product Family
Specifications
- Catalog No:
- GEHE17-1353-01
- GEHE17-1355-01
- Max. pressure:
- 5 bar
- 5 bar
- pH range:
- 3 - 13
- 3 - 13
Specifications
Product Family Options
Product Information
- Column hardwareParticle SizeBed dimensionsCapacityFlow rateAvailabilityPrice
- GEHE17-1353-01HiTrap™ phenyl FF (low sub)Specifications:
More
Specifications
Cat. No.GEHE17-1353-01Max. pressure5 barpH range3 - 13 - GEHE17-1355-01HiTrap™ phenyl FF (high sub)Specifications:
More
Specifications
Cat. No.GEHE17-1355-01Max. pressure5 barpH range3 - 13