About this item
VWR® offers a complete range of products for the determination of the hydrocarbon oil index (HOI) in accordance with EN ISO 9377-2 (DIN H53).
- All standards are CRMs according to ISO 17025 and ISO 17034
- Shelf life of 24 months
- Storage: 2 – 8°C
- Supplied with a detailed, batch-specific certificate of analysis
The Hydrocarbon Oil Index (HOI) is defined as the total amount of compounds which can be extracted from the sample (drinking water, surface water and waste water) with a non-polar solvent having a boiling point between 39°C and 69°C .
In addition, the compounds must not absorb on Florisil® (magnesium sulphate) and must elute between n-decane (C10H24) and n-tetracontane (C40H82) when analyzed by GC using an apolar analytical column.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- CAS No.:2778-96-3
- Formula:C₃₆H₇₂O₂
- Description:Stearyl stearate
Specifications
Related Information
Certificates
Search by Lot NumberSDS_EN_UK_VWRC87495.pdf SDS_EN_UK_VWRC88391.pdf SDS_EN_UK_VWRC88392.pdf SDS_EN_UK_VWRC88419.pdf sds_VWRC87495
SDS
VWR® for Calibrations