Order Entry
China
ContactUsLinkComponent
J.T.Baker® BAKER INSTRA-ANALYZED®, Silicon Standard
J.T.Baker® BAKER INSTRA-ANALYZED®, Silicon Standard
Catalog # BAKR6466-04
Supplier:  Avantor
CAS Number:  
J.T.Baker® BAKER INSTRA-ANALYZED®, Silicon Standard
Catalog # BAKR6466-04
Supplier:  Avantor
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pk:
    150 mL
  • Accuracy:
    ±10 mg/L
  • Element:
    Silicon
  • Symbol:
    Si

Specifications

About this item

Ammonium hexafluorosilicate in water.

  • Traceable to NIST