




Pyridine ≥99.0% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:110-86-1
:
Formula:C₅H₅N
MDL Number:MFCD00011732
Merck Index:13,08060
Boiling Pt:115 °C (1013 hPa)
Melting Pt:–44 °C
Density:0.983 g/cm³ (20 °C)
Flash Pt:17 °C
Storage Temperature:Ambient
EINECS:203-809-9
UN:1282
ADR:3,II
:Management of Change (MOC) category = N
:79.1 g/mol
:GHS02
:GHS05
:GHS07
:GHS08
:GHS09
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- ACS - Assay (C₅H₅N) (by GC, corrected for water):≥99.0 %
- ACS - Solubility in H₂O:Passes Test
- ACS - Residue after Evaporation:≤0.002 %
- ACS - Water (H₂O)(by Karl Fischer titrn):≤0.1 %
- ACS - Chloride (Cl):≤0.001 %
- ACS - Sulfate (SO₄):≤0.001 %
- ACS - Ammonia (as NH₃):≤0.002 %
- ACS - Copper (Cu):≤5 ppm
- ACS - Reducing Substances:Passes Test