Order Entry
China
ContactUsLinkComponent
 
Flammable
Irritant
Acetone, low water content ≥99.3%, BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Catalog # BAKR9003-03
Supplier:  Avantor
   Safer Human and Environmental Health
CAS Number:  
Flammable
Irritant
Acetone, low water content ≥99.3%, BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Catalog # BAKR9003-03
Supplier:  Avantor
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    4 L
  • Seq:
    5162
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-350 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:
    ≤330
  • Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:
    ≤0.3 ppb
  • Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:
    ≤1.0 ppb
  • Assay ((CH₃)₂CO)(by GC):
    ≥99.3 %
  • Residue after Evaporation:
    ≤2.0 ppm
  • Titrable Acid (µeq/g):
    ≤0.3
  • Titrable Base (µeq/g):
    ≤0.6
  • Water (H₂O):
    ≤0.10 %

Specification Test Results

About this item

 :
 :58.08 g/mol
Merck Index:13,00067
Boiling Pt:56.2 °C (1013 hPa)
Melting Pt:–95.4 °C
Density:0.792 g/cm³ (20 °C)
Flash Pt:<–20 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
 :Management of Change (MOC) category = NI
REACH:01-2119471330-49
 :GHS02
 :GHS07
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765