Order Entry
China
Orders LinkContactUsLinkComponent
Thread adaptors TYPE C, b.safe
Thread adaptors TYPE C, b.safe
Supplier:   BOHLENDER
Thread adaptors TYPE C, b.safe
Supplier:  BOHLENDER
Certificates
Thread adaptors TYPE C, b.safe
Thread adaptors TYPE C, b.safe
6 Options Available Below
Product Details & Documents
All adaptors of this type are supplied with two female threads and recessed grips.
  • Stay flexible and independent
  • With recessed grips for an easy assembly
Your existing caps and waste caps do not fit with the flasks and canisters which you would like to use or vice versa? Simply use b.safe thread adaptors for a flexible combination of caps and waste caps independently from the thread on the flask and/or container.
Product Family Options
Product Information
  • Material
    Thread
    Availability
    Price
  • PE
    Tri-Sure 2" (female), S 60/61 (female)
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • PP
    G 2"/BSP 2" (female), S 55 (female)
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • PP
    G 2"/BSP 2" (female), S 60/61 (female)
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • PP
    G 2"/BSP 2" (female), S 65 (female)
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • PP
    G 2"/BSP 2" (female), S 70/71 (female)
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • PP
    G 1¹/₂" (female), GL 45 (female)
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件