Human recombinant integrin alpha 4 beta 7 (from HEK293)
ACRBIT7-H52W4-1MG
:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Pk:1 mg
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Seq:02
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised Natalizumab at 1 μg/ml (100 μl/well) can bind Human ITGA4 and ITGB7 Heterodimer Protein, His Tag and Tag Free (Cat. No. ACRBIT7-H52W4) with a linear range of 4 - 63 ng/ml (QC tested).
- Gene ID:P13612-1 (ITGA4) and P26010-1 (ITGB7)
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:ITGA4&ITGB7|Integrin alpha 4 beta 7
- Protein/peptide name:Integrin alpha 4 beta 7
- Purity:>90%
- Molecular weight:111,4 kDa (ITGA4) and 81,8 kDa (ITGB7)
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT