- Description:ASSIST PLUS pipetting robot
- Pk:1
INTEGRA has developed the ASSIST PLUS pipetting robot to streamline routine pipetting tasks. Using INTEGRA electronic multichannel pipettes, the system automates pipetting tasks, eliminates physical strain and ensures superior reproducibility and error free pipetting.
- Automates any VOYAGER or VIAFLO multichannel electronic pipette
- Compatible with all INTEGRA tube racks, reagent reservoirs and microplates (12-384 well)
- Protocols are created either on the pipette or in VIALAB (free for download)
- Easy and fast programming with the VIALAB pipetting software
Simply mount an INTEGRA VIAFLO or VOYAGER electronic pipette onto the ASSIST PLUS pipette holder, choose a pipetting protocol, add a tip rack and your labware. Press RUN and ASSIST PLUS will automatically carry out the pipetting task. ASSIST PLUS can be used for serial dilutions, reformatting, plate filling and reagent addition and it’s possible to add options of heating, cooling and shaking of the labware. For setting up the protocol you use VIALAB – an intuitive computer program where you can visualize the protocol before running it on the ASSIST PLUS.