Order Entry
China
ContactUsLinkComponent
VWR® ARISTAR®, Osmium Standard Solution for ICP
VWR® ARISTAR®, Osmium Standard Solution for ICP
Catalog # VWRC87051.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Osmium Standard Solution for ICP
Catalog # VWRC87051.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HCl
  • Pk:
    100 mL
  • Accuracy:
    ±0,5 mg/L
  • Element:
    Osmium
  • Symbol:
    Os

Specifications

About this item

  • Produced from high purity OsCl₃
  • Certified reference material
  • Traceable to NIST
  • Certified Value and uncertainty 1000,0 ± 0,5 mg/L

Density 1,060 g/cm³ at 20 °C.

Caution: Storage at ambient temperatures.