Order Entry
China
ContactUsLinkComponent
Cleanroom gloves, latex, Kimtech Pure* G5
Cleanroom gloves, latex, Kimtech Pure* G5
  • Catalog No:
  • KIMB56854
  • KIMB56853
  • KIMB56855
  • KIMB56852
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex
  • Latex
  • Latex
  • Latex
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Cuff thickness:
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • 0,16 mm
  • Finger thickness:
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • Palm thickness:
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • Tensile strength:
  • 24 MPa, min (before aging)
    18 MPa, min.(after aging)
  • 24 MPa, min (before aging)
    18 MPa, min.(after aging)
  • 24 MPa, min (before aging)
    18 MPa, min.(after aging)
  • 24 MPa, min (before aging)
    18 MPa, min.(after aging)