
Citric acid kit, anhydrous 99.5-100.5% (by anhydrous basis), powder USP, J.T.Baker®
Catalog # BAKR0122-K1
Supplier: Avantor
CAS Number:
Specifications
- Pk:1 KIT
- Seq:0075
- Pk Info:3971 g + 200 g
Specifications
Specification Test Results
- Kit Components Meet Requirements:Passes Test
Specification Test Results
About this item
:
:192.13 g/mol
Merck Index:13,02350
Boiling Pt:310 °C (1013 hPa)
Melting Pt:149…151 °C
Density:1.44 g/cm³
Storage Temperature:Ambient
EINECS:201-069-1
REACH:01-2119457026-42-0000
:GHS07
CAS Number:77-92-9
Formula:HOC(COOH)(CH₂COOH)₂
MDL Number:MFCD00011669