订单输入
China
ContactUsLinkComponent
VWR® Adhesive Sealing Strips, PP, 1×8
VWR® Adhesive Sealing Strips, PP, 1×8
Catalog # VWRI391-0639
Supplier:  VWR Collection
undefined
VWR® Adhesive Sealing Strips, PP, 1×8
Catalog # VWRI391-0639
Supplier:  VWR Collection

Specifications

  • Optical property:
    Transparent
  • 灭菌:
    -
  • 说明:
    1×8 sealing strips
  • 厚度:
    74,9 µm
  • 包装规格:
    400

Specifications

About this item

1x8 adhesive sealing strips are ideal for use with 1x8 strip-well ELISA plates.

  • Ideal for ELISA, incubation and general use
  • Seals a single 8-well row of a 96-well plate or an ELISA strip-well plate
  • 2,0 mil PP film
  • Compatible with both polystyrene and polypropylene plates
  • Certified DNase- and RNase-free
  • Recommended temperature range from −40 to +120 °C

8 strips are pre-cut on a continuous liner for ease of use.