Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% USP, Multi-Compendial, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:77-86-1
:
Formula:NH₂C(CH₂OH)₃
MDL Number:MFCD00004679
:121.14 g/mol
:Management of Change (MOC) category = R
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- USP - Assay (C₄H₁₁NO₃):99.0 - 101.0 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Identification C:Passes Test
- USP - pH (1 in 20):10.0 - 11.5
- USP - Loss on Drying:≤1.0 %
- USP - Residue on Ignition:≤0.1 %
- USP - Melting Range(Lower):168 - 172 °C
- USP - Melting Range(Upper):168 - 172 °C
- EP/BP - Assay (C₄H₁₁NO₃) (dried basis):99.0 - 100.5 %
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - pH:10.0 - 11.5
- EP/BP - Related Substances:≤1.0 %
- EP/BP - Chloride (Cl):≤100 ppm
- EP/BP - Iron (Fe):≤10 ppm
- EP/BP - Loss on Drying at 105°C:≤0.5 %
- EP/BP - Ash (sulfated):≤0.1 %
- JPC 1997 - Assay (C₄H₁₁NO₃) (dried basis):≥99.0 %
- JPC 1997 - Identification A:Passes Test
- JPC 1997 - Identification B:Passes Test
- JPC 1997 - pH:10.3 - 10.7
- JPC 1997 - Melting Point:168 - 172 °C
- JP - Clarity and Color of Solution:Passes Test
- JPC 1997 - Heavy Metals (as Pb):≤8 ppm
- JPC 1997 - Arsenic (As):≤1.6 ppm
- JPC 1997 - Loss on Drying:≤0.5 %
- JPC 1997 - Residue on Ignition:≤0.10 %
- Assay (C₄H₁₁NO₃) (dried basis):≥99.9 %
- Absorbance of a 1M Solution - 260 nm:≤0.06
- Absorbance of a 1M Solution - 280 nm:≤0.06
- Absorbance of a 1M Solution - 400 nm:≤0.01
- Endotoxin Concentration (EU/g):≤2.5
- Iron (Fe):≤1 ppm
- Lead (Pb):≤1 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_BAKRLP12-10 spec_BAKRLP12-D1 spec_BAKRLP12-S1