L(+)-Arginine 98.5-101.5% (dry basis) USP, Multi-Compendial, J.T.Baker®
: Avantor
:74-79-3
:
:C₆H₁₄N₄O₂
:MFCD00002635
:174.2 g/mol
:Management of Change (MOC) category = R
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- USP - Assay (C₆H₁₄N₄O₂) (dried basis):98.5 - 101.5 %
- USP - Identification:Passes Test
- USP - Specific Rotation [Å]²⁵ ^D (+):26.3 - 27.7 Degree
- USP - Loss on Drying at 105°C:≤ 0.5 %
- USP - Chloride (Cl):≤ 0.05 %
- USP - Residue on Ignition:≤ 0.3 %
- USP - Sulfate (SO₄):≤ 0.03 %
- USP - Iron (Fe):≤ 0.003 %
- USP - Related Compounds - Individual Impurities:≤ 0.5 %
- USP - Related Compounds - Total Impurities:≤ 2.0 %
- EP/BP - Assay (C₆H₁₄N₄O₂) (dried basis):98.5 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Specific Rotation [Å]²º^D (+):25.5 - 28.5 Degree
- EP/BP-Ninhydrin-Positive Substances-Each:≤ 0.2 %
- EP/BP-Ninhydrin-Positive Substances-Total Impurities:≤ 0.5 %
- EP/BP - Chloride (Cl):≤ 200 ppm
- EP/BP - Sulfate (SO₄):≤ 300 ppm
- EP/BP - Ammonium (NH₄):≤ 0.02 %
- EP/BP - Iron (Fe):≤ 10 ppm
- EP/BP - Loss on Drying:≤ 0.5 %
- EP/BP - Ash (sulfated):≤ 0.1 %
- JP - Identification:Passes Test
- JP - Optical rotation, 20° C (+):26.9 - 27.9 Degree
- JP - pH (1:10):10.5 - 12.0
- JP - Clarity and Color of Solution:Passes Test
- JP - Chloride (Cl):≤ 0.021 %
- JP - Sulfate (SO₄):≤ 0.028 %
- JP - Ammonium (NH₄):≤ 0.02 %
- JP - Heavy Metals (as Pb):≤ 10 ppm
- JP - Iron (Fe):≤ 10 ppm
- JP - Related Substances:Passes Test
- JP - Loss on Drying at 105°C:≤ 0.30 %
- JP - Residue after Ignition:≤ 0.1 %
- JP - Assay (C₆H₁₄N₄O₂)(dried basis):98.5 - 101.0 %