Dextrose, anhydrous 97.5-102.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:50-99-7
:492-62-6
Formula:C₆H₁₂O₆
MDL Number:MFCD00148912
:Management of Change (MOC) category = R
:180.16 g/mol
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Identification C: Water (H₂O):≤1.0 %
- USP - Assay (Anhydrous basis):97.5 - 102.0 %
- USP - Related substances: Maltose and isomaltose:≤0.4 %
- USP - Related substances: Maltotriose:≤0.2 %
- USP - Related substances: Fructose:≤0.15 %
- USP - Related substances: Unspecified impurities, each:≤0.10 %
- USP - Related substances: Total Impurities:≤0.5 %
- USP - Color and Clarity of Solution:Passes Test
- USP - Dextrin:Passes Test
- USP - Soluble Starch, Sulfites:≤15 ppm
- USP - Conductivity (uS cm-1):≤20 µS/cm
- USP - Endotoxin Concentration (EU/g):≤2.5
- EP/BP - Identification A (Specific Rotation (+)):52.5 - 53.3 Degree
- EP/BP - Identification B:Passes Test
- EP/BP - Identification E:Passes Test
- EP/BP - Assay (C₆H₁₂O₆) (anhydrous substance):97.5 - 102.0 %
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Conductivity, uS cm-1:≤20
- EP/BP - Related Substances: sum of impurities A and B:≤0.4 %
- EP/BP - Related Substances: impurity C:≤0.2 %
- EP/BP - Related Substances: impurity D:≤0.15 %
- EP/BP - Related Substances: unspecified impurities, each:≤0.10 %
- EP/BP - Related Substances: total impurities:≤0.5 %
- EP/BP - Dextrins:Passes Test
- EP/BP - Soluble starch, sulfite:≤15 ppm
- EP/BP - Water (H₂O):≤1.0 %
- JP - Identification 1:Passes Test
- JP - Identification 2:Passes Test
- JP - Water (H₂O):≤1.0 %
- JP - Assay (Anhydrous basis):97.5 - 102.0 %
- JP - Maltose and isomaltose:≤0.4 %
- JP - Maltotriose:≤0.2 %
- JP - Fructose:≤0.15 %
- JP - Unspecified Impurities, each:≤0.10 %
- JP - Total Impurities:≤0.5 %
- JP - Color and Clarity of Solution:Passes Test
- JP - Soluble Starch, Sulfites:≤15 ppm
- JP - Conductivity (uS cm-1):≤20 µS/cm
- JP - Dextrin:Passes Test
- JP - Heavy Metals (as Pb):≤4 ppm