Sodium acetate trihydrate 99.0-101.0% (dry basis), crystals USP, Multi-Compendial, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:6131-90-4
:
Formula:H₃CCOONa·3H₂O
MDL Number:MFCD00071557
:136.08 g/mol
:Management of Change (MOC) category = HR
Flash Pt:250.0 °C
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- USP - Assay (CH₃COONa) (dried basis):99.0 - 101.0 %
- USP - Calcium and Magnesium:Passes Test
- USP - Chloride (Cl):≤ 0.035 %
- USP - Endotoxin Concentration (EU/g):≤ 2.5
- USP - Identification:Passes Test
- USP - Insoluble Matter:≤ 0.05 %
- USP - Loss on Drying:38.0 - 41.0 %
- USP - pH of 5% Solution at 25°C:7.5 - 9.2
- USP - Potassium (K):Passes Test
- USP - Sulfate (SO₄):≤ 0.005 %
- EP/BP - Assay (CH₃COONa):99.0 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - pH (1 in 20):7.5 - 9.0
- EP/BP - Reducing Substances:Passes Test
- EP/BP - Chloride (Cl):≤ 200 ppm
- EP/BP - Sulfate (SO₄):≤ 200 ppm
- EP/BP - Calcium and Magnesium:≤ 50 ppm
- EP/BP - Iron (Fe):≤ 10 ppm
- EP/BP - Loss on Drying:39.0 - 40.5 %
- JP - Assay (CH₃COONa):99.5 - 101.0 %
- JP - Identification:Passes Test
- JP - Clarity and Color of Solution:Passes Test
- JP - Acidity or Alkalinity:Passes Test
- JP - Chloride (Cl):≤ 0.011 %
- JP - Sulfate (SO₄):≤ 0.017 %
- JP - Heavy Metals (as Pb):≤ 10 ppm
- JP - Calcium and Magnesium:Passes Test
- JP - Arsenic (As):≤ 2 ppm
- JP - Substances Reducing Permanganate:Passes Test
- JP - Loss on Drying:39.0 - 40.5 %
- FCC - Alkalinity:≤ 0.05 %
- FCC - Assay (CH₃COONa) (dried basis):99.0 - 101.0 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Lead (Pb):≤ 2 mg/kg
- FCC - Loss on Drying:38.0 - 41.0 %
- FCC - Potassium Compounds:Passes Test