Order Entry
China
ContactUsLinkComponent
Sodium dihydrogen phosphate monohydrate 98.0-103.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Sodium dihydrogen phosphate monohydrate 98.0-103.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Sodium dihydrogen phosphate monohydrate 98.0-103.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specification Test Results

  • Appearance (White crystals or granules):
    Passes Test
  • USP - Assay (anhydrous basis):
    98.0 - 103.0 %
  • USP - Aluminum, Calcium, and Related Elements:
    Passes Test
  • USP - Arsenic (As):
    ≤ 8 ppm
  • USP - Chloride (Cl):
    ≤ 0.014 %
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Insoluble Substances:
    ≤ 0.2 %
  • USP - pH (1 in 20):
    4.1 - 4.5
  • USP - Sulfate (SO₄):
    ≤ 0.15 %
  • USP - Water (H₂O):
    10.0 - 15.0 %
  • EP - Identification A:
    Passes Test
  • EP - Identification B:
    Passes Test
  • EP - Identification C:
    Passes Test
  • EP - Assay (anhydrous basis):
    98.0 - 100.5 %
  • EP - Chloride (Cl):
    ≤ 200 ppm
  • EP - Sulfate (SO₄):
    ≤ 300 ppm
  • EP - Reducing Substances:
    Passes Test
  • Endotoxin Concentration (EU/g):
    ≤ 2.5
  • EP - Iron (Fe):
    ≤ 10 ppm
  • EP - Appearance of Solution:
    Passes Test
  • EP - pH:
    4.2 - 4.5
  • BP - Identification A:
    Passes Test
  • BP - Identification B:
    Passes Test
  • BP - Identification C:
    Passes Test
  • BP - Identification D:
    Passes Test
  • BP - Acidity:
    4.2 - 4.5
  • BP - Clarity and Color of Solution:
    Passes Test
  • BP - Arsenic (As):
    ≤ 2 ppm
  • BP - Iron (Fe):
    ≤ 10 ppm
  • BP - Chloride (Cl):
    ≤ 200 ppm
  • BP - Sulfate (SO₄):
    ≤ 300 ppm
  • BP - Reducing Substances:
    Passes Test
  • BP - Loss on Drying at 130°C:
    11.5 - 14.5 %
  • BP - Assay (dried basis):
    98.0 - 100.5 %
  • JPE - Assay (dried basis):
    ≥ 98.0 %
  • JPE - Identification:
    Passes Test
  • JPE - pH:
    4.1 - 4.7
  • JPE - Clarity and Color of Solution:
    Passes Test
  • JPE - Chloride (Cl):
    ≤ 0.005 %
  • JPE - Sulfate (SO4):
    ≤ 0.019 %
  • JPE - Heavy Metals:
    ≤ 10 ppm
  • JPE - Arsenic (As):
    ≤ 2 ppm

Specification Test Results

Related Information