L-Methionine 98.5-101.5% (dry basis) USP, Multi-Compendial, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:63-68-3
:
Formula:C₅H₁₁NO₂S
MDL Number:MFCD00063097
:149.21 g/mol
Melting Pt:280…282 °C
:Management of Change (MOC) category = R
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- USP - Assay (C₅H₁₁NO₂S) (dried basis):98.5 - 101.5 %
- USP - Identification:Passes Test
- USP - Specific Rotation [Å]²⁵ ^D (+):22.4 - 24.7 Degree
- USP - pH:5.6 - 6.1
- USP - Loss on Drying at 105°C:≤0.3 %
- USP - Residue on Ignition:≤0.4 %
- USP - Chloride (Cl):≤0.05 %
- USP - Sulfate (SO₄):≤0.03 %
- USP - Iron (Fe):≤0.003 %
- USP - Related Compounds - Methionine sulfoxide:≤0.1 %
- USP - Related Compounds - N-Acetyl-D,L- methionine:≤0.2 %
- USP - Related Compounds - Any unspecified impurity:≤0.1 %
- USP - Related Compounds - Total unspecified impurities:≤0.30 %
- EP/BP - Assay (C₅H₁₁NO₂S) (dried basis):99.0 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - pH:5.5 - 6.5
- EP/BP - Specific Rotation [Å]²º^D (+):22.5 - 24.0 Degree
- EP/BP - Related Substances: Unspecified Impurities, each:≤0.10 %
- EP/BP - Related Substances: Total Impurities:≤0.3 %
- EP/BP - Chloride (Cl):≤200 ppm
- EP/BP - Sulfate (SO₄):≤300 ppm
- EP/BP - Ammonium (NH₄):≤200 ppm
- EP/BP - Iron (Fe):≤10 ppm
- EP/BP - Loss on Drying at 105°C:≤0.5 %
- EP/BP - Ash (sulfated):≤0.1 %
- JP - Assay (C₅H₁₁NO₂S) (dried basis):98.5 - 101.0 %
- JP - Identification:Passes Test
- JP - Optical Rotation (+):21.0 - 25.0 Degree
- JP - pH:5.2 - 6.2
- JP - Clarity and Color of Solution:Passes Test
- JP - Chloride (Cl):≤0.021 %
- JP - Sulfate (SO₄):≤0.028 %
- JP - Ammonium (NH₄):≤0.02 %
- JP - Heavy Metals (as Pb):≤20 ppm
- JP - Arsenic (As):≤2 ppm
- JP - Related Substances:Passes Test
- JP - Loss on Drying at 105°C:≤0.30 %
- JP - Residue after Ignition:≤0.10 %
- Bacterial endotoxins, EU/g:≤100
- Total Aerobic Microbial Count:≤100 cfu/g
- Total Yeasts and Molds:≤100 cfu/g