Order Entry
China
ContactUsLinkComponent
 
Irritant
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.0% (dried basis), Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
Catalog # BAKR4108-07
Supplier:  Avantor
CAS Number:  
undefined
Irritant
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.0% (dried basis), Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
Catalog # BAKR4108-07
Supplier:  Avantor
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    PE pail, screw top
  • Pk:
    12 kg
  • Seq:
    4187
  • Pk Info:
    1×12 kg

Specifications

Specification Test Results

  • Assay (NH₂C(CH₂OH)₃·HCl) (dried basis):
    ≥99.0 %
  • Appearance:
    Passes Test
  • Identification (by IR):
    Passes Test
  • Absorbance of a 1M Solution - 260 nm:
    ≤0.06
  • Absorbance of a 1M Solution - 280 nm:
    ≤0.06
  • Absorbance of a 1M Solution - 400 nm:
    ≤0.01
  • Water (H₂O)(by Karl Fischer titrn):
    ≤0.5 %
  • RNase Activity (None Detected):
    Passes Test
  • DNase Activity (None Detected):
    Passes Test
  • Protease Activity (None Detected):
    Passes Test
  • Trace Impurities - Calcium (Ca):
    ≤1 ppm
  • Trace Impurities - Copper (Cu):
    ≤1 ppm
  • Trace Impurities - Iron (Fe):
    ≤1 ppm
  • Trace Impurities - Lead (Pb):
    ≤1 ppm
  • Trace Impurities - Magnesium (Mg):
    ≤1.000 ppm

Specification Test Results

About this item

 :
 :157.6 g/mol
 :Management of Change (MOC) category = R
 :GHS07
CAS Number:1185-53-1
Formula:NH₂C(CH₂OH)₃·HCl
MDL Number:MFCD00012590