
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.0% Made from Tromethamine USP, Biotech Reagent, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:1185-53-1
:
Formula:NH₂C(CH₂OH)₃·HCl
MDL Number:MFCD00012590
:157.6 g/mol
Merck Index:12,09902
Melting Pt:147…153 °C
Density:1.05 g/ml (20 °C)
:Management of Change (MOC) category = R
EINECS:214-684-5
:GHS07
Storage Temperature:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Assay (NH₂C(CH₂OH)₃·HCl):≥99.0 %
- Appearance (colorless crystals to white crystalline powder):Passes Test
- Absorbance of a 1M Solution - 260 nm:≤0.06
- Absorbance of a 1M Solution - 280 nm:≤0.06
- Absorbance of a 1M Solution - 400 nm:≤0.01
- Identification (by IR):Passes Test
- Loss on Drying at 105°C:≤0.5 %
- Endotoxin Concentration (EU/g):≤2.5
- Melting Point:147 - 153 °C
- pH of 0.5M Solution at 25°C:3.5 - 5.0
- Trace Impurities - Arsenic (As):≤10 ppm
- Trace Impurities - Calcium (Ca):≤10.0 ppm
- Trace Impurities - Copper (Cu):≤10 ppm
- Trace Impurities - Iron (Fe):≤10 ppm
- Trace Impurities - Lead (Pb):≤10 ppm
- Trace Impurities - Magnesium (Mg):≤10 ppm