
Sodium chloride 99.0-100.5% (dried basis), granular USP, Multi-Compendial, J.T.Baker®
Catalog # BAKR3625-07
Supplier: Avantor
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:HDPE pail, Flowmor™
- Pk:12 kg
- Seq:4187
- Pk Info:1×12 kg
Specifications
Specification Test Results
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Acidity or Alkalinity:Passes Test
- USP - Appearance of Solution:Passes Test
- USP - Aluminum (Al):≤0.2 ppm
- USP - Arsenic (As):≤1 ppm
- USP - Barium (Ba):Passes Test
- USP - Bromide (Br):≤100 ppm
- USP - Ferrocyanide:Passes Test
- USP - Iodides (I):Passes Test
- USP - Iron (Fe):≤2 ppm
- USP - Loss on Drying at 105°C:≤0.5 %
- USP - Magnesium and Alkaline Earth Metals (as Ca):≤100 ppm
- USP - Nitrite (NO₂) (Absorbance Value):≤0.01
- USP - Phosphate (PO₄):≤25 ppm
- USP - Potassium (K):≤500 ppm
- USP - Sulfate (SO₄):≤200 ppm
- USP - Assay (NaCl) (dried basis):99.0 - 100.5 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Aluminum (Al):≤0.2 ppm
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Acidity or Alkalinity:Passes Test
- EP/BP - Bromide (Br):≤100 ppm
- EP/BP - Ferrocyanide:Passes Test
- EP/BP - Iodide (I):Passes Test
- EP/BP - Nitrite (NO₂):≤0.01
- EP/BP - Phosphate (PO₄):≤25 ppm
- EP/BP - Sulfate (SO₄):≤200 ppm
- EP/BP - Arsenic (As):≤1 ppm
- EP/BP - Barium (Ba):Passes Test
- EP/BP - Iron (Fe):≤2 ppm
- EP/BP - Magnesium and Alkaline Earth Metals:≤100 ppm
- EP/BP - Loss on Drying at 105°C:≤0.5 %
- EP/BP - Potassium (K):≤500 ppm
- EP/BP - Assay (NaCl) (dried basis):99.0 - 100.5 %
- EP/BP - Endotoxin Concentration, IU/g:≤2.5
- JP - Assay (NaCl) (dried basis):99.0 - 100.5 %
- JP - Identification A:Passes Test
- JP - Identification B:Passes Test
- JP - Acidity or Alkalinity:Passes Test
- JP - Clarity and Color of Solution:Passes Test
- JP - Bromide (Br):Passes Test
- JP - Ferrocyanide:Passes Test
- JP - Iodide (I):Passes Test
- JP - Iron (Fe):Passes Test
- JP - Magnesium and Alkaline Earth Metals:Passes Test
- JP - Phosphate (PO₄):Passes Test
- JP - Sulfates (SO₄):Passes Test
- JP - Heavy Metals (as Pb):≤3 ppm
- JP - Barium (Ba):Passes Test
- JP - Arsenic (As):≤2 ppm
- JP - Loss on Drying:≤0.5 %
- Iron (Fe):≤2 ppm
- Total Transition Metals (Cr, Co, Cu, Mn):≤10 ppm
Specification Test Results
About this item
:
:58.44 g/mol
:Management of Change (MOC) category = R
:GHS07
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477