


1,2-Dichloroethane ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:107-06-2
:
Formula:ClH₂CCH₂Cl
MDL Number:MFCD00000963
:98.96 g/mol
Density:1.254 g/cm³ (20 °C)
Flash Pt:13 °C
EINECS:203-458-1
UN:1184
ADR:3 (6.1),II
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Assay (GC):min. 99,8 %
- Water (KF):max. 0,01 %
- Titrable Acid (meq/g):max. 0,0005
- Residue after Evaporation :max. 0,0005 %
- 240 nm:max. 0,3
- 250 nm:max. 0,07
- 270 nm:max. 0,009
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC85923.290 spec_VWRC85923.290 spec_VWRC85923.320 spec_VWRC85923.320
Product specification
VWR® HiperSolv® Chromanorm® HPLC Reagents