Order Entry
China
ContactUsLinkComponent
Hydrogen peroxide 30% stabilised, CMOS for microelectronic use, J.T.Baker®
Hydrogen peroxide 30% stabilised, CMOS for microelectronic use, J.T.Baker®
Supplier:  Avantor
Certificates
Hydrogen peroxide 30% stabilised, CMOS for microelectronic use, J.T.Baker®
Supplier:  Avantor
Certificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (H₂O₂):
    31.52 - 31.72 %
  • Free Acid (µeq/g):
    ≤0.2
  • Residue after Evaporation:
    ≤10 ppm
  • Ammonium (NH₄):
    ≤3 ppm
  • Chloride (Cl):
    ≤0.2 ppm
  • Nitrate (NO₃):
    ≤2 ppm
  • Phosphate (PO₄):
    ≤1 ppm
  • Sulfate (SO₄):
    ≤3 ppm
  • Trace Impurities - Aluminum (Al):
    ≤47.550 ppb
  • Trace Impurities - Antimony (Sb):
    ≤10.000 ppb
  • Trace Impurities - Arsenic (As):
    ≤10.000 ppb
  • Arsenic and Antimony (as As):
    ≤10.0 ppb
  • Trace Impurities - Barium (Ba):
    ≤6.870 ppb
  • Trace Impurities - Beryllium (Be):
    ≤10.000 ppb
  • Trace Impurities - Bismuth (Bi):
    ≤20.000 ppb
  • Trace Impurities - Boron (B):
    ≤6.200 ppb
  • Trace Impurities - Cadmium (Cd):
    ≤10.000 ppb
  • Trace Impurities - Calcium (Ca):
    ≤10.080 ppb
  • Trace Impurities - Chromium (Cr):
    ≤4.930 ppb
  • Trace Impurities - Cobalt (Co):
    ≤2.000 ppb
  • Trace Impurities - Copper (Cu):
    ≤1.680 ppb
  • Trace Impurities - Gallium (Ga):
    ≤20.000 ppb
  • Trace Impurities - Germanium (Ge):
    ≤10.000 ppb
  • Trace Impurities - Gold (Au):
    ≤10.000 ppb
  • Heavy Metals (as Pb):
    ≤500.0 ppb
  • Trace Impurities - Iron (Fe):
    ≤13.470 ppb
  • Trace Impurities - Lead (Pb):
    ≤2.430 ppb
  • Trace Impurities - Lithium (Li):
    ≤1.920 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤5.230 ppb
  • Trace Impurities - Manganese (Mn):
    ≤1.560 ppb
  • Trace Impurities - Molybdenum (Mo):
    ≤10.000 ppb
  • Trace Impurities - Nickel (Ni):
    ≤3.260 ppb
  • Trace Impurities - Niobium (Nb):
    ≤10.000 ppb
  • Trace Impurities - Potassium (K):
    ≤850.000 ppb
  • Trace Impurities - Silicon (Si):
    ≤100.000 ppb
  • Trace Impurities - Silver (Ag):
    ≤10.000 ppb
  • Trace Impurities - Sodium (Na):
    ≤39.700 ppb
  • Trace Impurities - Strontium (Sr):
    ≤10.000 ppb
  • Trace Impurities - Tantalum (Ta):
    ≤10.000 ppb
  • Color (APHA):
    ≤10
  • Particle count at point of fill - 0.1 µm and higher (RION KS42A):
    ≤23129.3 par/ml
  • Particle count at point of fill - 0.2 µm and higher (RION KS42A):
    ≤1596.8 par/ml
  • Particle count at point of fill - 0.5 µm and higher (RION KS42A):
    ≤350.0 par/ml

Specification Test Results

Related Information

Certificates
Search by Lot Number