Sodium chloride 99.0-100.5% (dried basis), granular USP, Multi-Compendial
Catalog # BAKR3627-L1
Supplier: Avantor
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pk:1 KIT
- Seq:0075
- Environmentally Preferable:
- Pk Info:1× 100 kg Kit/Bag
Specifications
Specification Test Results
- Kit Components Meet Requirements:Passes Test
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Acidity or Alkalinity:Passes Test
- USP - Appearance of Solution:Passes Test
- USP - Aluminum (Al):≤0.2 ppm
- USP - Arsenic (As):≤1 ppm
- USP - Barium (Ba):Passes Test
- USP - Bromide (Br):≤100 ppm
- USP - Ferrocyanide:Passes Test
- USP - Iodides (I):Passes Test
- USP - Iron (Fe):≤2 ppm
- USP - Loss on Drying at 105°C:≤0.5 %
- USP - Magnesium and Alkaline Earth Metals (as Ca):≤100 ppm
- USP - Nitrite (NO₂) (Absorbance Value):≤0.01
- USP - Phosphate (PO₄):≤25 ppm
- USP - Potassium (K):≤500 ppm
- USP - Sulfate (SO₄):≤200 ppm
- USP - Assay (NaCl) (dried basis):99.0 - 100.5 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Aluminum (Al):≤0.2 ppm
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Acidity or Alkalinity:Passes Test
- EP/BP - Bromide (Br):≤100 ppm
- EP/BP - Ferrocyanide:Passes Test
- EP/BP - Iodide (I):Passes Test
- EP/BP - Nitrite (NO₂):≤0.01
- EP/BP - Phosphate (PO₄):≤25 ppm
- EP/BP - Sulfate (SO₄):≤200 ppm
- EP/BP - Arsenic (As):≤1 ppm
- EP/BP - Barium (Ba):Passes Test
- EP/BP - Iron (Fe):≤2 ppm
- EP/BP - Magnesium and Alkaline Earth Metals:≤100 ppm
- EP/BP - Loss on Drying at 105°C:≤0.5 %
- EP/BP - Potassium (K):≤500 ppm
- EP/BP - Assay (NaCl) (dried basis):99.0 - 100.5 %
- EP/BP - Endotoxin Concentration, IU/g:≤2.5
- JP - Assay (NaCl) (dried basis):99.0 - 100.5 %
- JP - Identification A:Passes Test
- JP - Identification B:Passes Test
- JP - Acidity or Alkalinity:Passes Test
- JP - Clarity and Color of Solution:Passes Test
- JP - Bromide (Br):Passes Test
- JP - Ferrocyanide:Passes Test
- JP - Iodide (I):Passes Test
- JP - Iron (Fe):Passes Test
- JP - Magnesium and Alkaline Earth Metals:Passes Test
- JP - Phosphate (PO₄):Passes Test
- JP - Sulfates (SO₄):Passes Test
- JP - Heavy Metals (as Pb):≤3 ppm
- JP - Barium (Ba):Passes Test
- JP - Arsenic (As):≤2 ppm
- JP - Loss on Drying:≤0.5 %
Specification Test Results
About this item
Synonyms: Saline, NaCl, Sea salt, Halite
:
:58.44 g/mol
Merck Index:14,08599
Boiling Pt:1413 °C (1013 hPa)
Melting Pt:801 °C
Density:2.16 g/cm³ (20 °C)
:Management of Change (MOC) category = R
Storage Temperature:Ambient
EINECS:231-598-3
:GHS07
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477