Order Entry
China
ContactUsLinkComponent
VWR®, Microscope Slides, Plain and Frosted
VWR®, Microscope Slides, Plain and Frosted
Catalog # VWRU48300-026
undefined
VWR®, Microscope Slides, Plain and Frosted
Catalog # VWRU48300-026
Specifications
  • Description:
    Frosted slides
  • L×W:
    75×25 mm
  • Material:
    Glass
  • Seq:
    01
  • Thickness:
    1,0 mm
  • Pk:
    72
Specifications
About this item
These pre-cleaned slides are made from high quality glass.
  • Glass fabrication
Frosted slides are sandblasted on one end of one side for marking specimens.
Double frosted slides are sandblasted on one end of both sides.
Economy slides are made from a lesser-quality sheet glass.