Order Entry
China
ContactUsLinkComponent
VWR®, Sealing Film Roll with Pre-Scored Sheets
VWR®, Sealing Film Roll with Pre-Scored Sheets
Catalog # VWRU76293-222
undefined
VWR®, Sealing Film Roll with Pre-Scored Sheets
Catalog # VWRU76293-222

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Packed:
    500 sheets/roll
  • Sterile:
    -
  • Description:
    Sealing film roll
  • Material:
    Polyester
  • Thickness:
    51 µm
  • Pk:
    1

Specifications

About this item

An economical, efficient way to rapidly seal microplates; faster than applying single sheeted film.

  • Acrylic adhesive
  • Dimension: 79,4×121,9 mm
  • Temperature range: –40...120 °C (–40...248 °F)
  • Ideal for ELISA, incubation, and general use
  • A self-standing, non-slip dispenser is supplied for ease of use

It is useful as a simple cover to protect samples from external contamination, dust, and debris.

2 mil polyester film minimises evaporation, reduces cross-contamination, and prevents spillage.