Order Entry
China
ContactUsLinkComponent
 
VWR®, Burette, Straight Bore PTFE Stopcock, Class A, Serialised
  VWRU75997-910
undefined
VWR®, Burette, Straight Bore PTFE Stopcock, Class A, Serialised
  VWRU75997-910

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Capacity:
    50 ml
  • Division:
    0,1 ml
  • Pk:
    1
  • Tolerance:
    ±0,05 ml

 

 

VWR® buret is made from borosilicate 3.3 glass, ASTM 438 Type 1, class A standard.

  • With external Ø of 13 mm
  • Supplied with a chemically-resistant, self-lubricating PTFE stopcock plug
  • Used primarily for titration, to deliver one reactant until the precise endpoint is reached
  • Calibration and tolerances per ASTM E287, ASTM E542, ASTM E694, and ISO 385 Standards

The graduations are typically calibrated 'to deliver' (marked "TD" or "EX") at 20 to 27 °C and indicated correspondingly on a label. Serialised, individually certified products.

These buret are calibrated to deliver a precise volume at a particular temperature.